2-Chlornicotinamid

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS22366G100 AS22366G25
ACTIAS22366G100EA 127 CHF
ACTIAS22366G100 ACTIAS22366G25
2-Chlornicotinamid
2-Chlornicotinamid
Formel: C₆H₅ClN₂O
MW: 156,57 g/mol
Melting Pt: 164…165 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006237
CAS-Nummer: 10366-35-5
EINECS: 233-808-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR