3-Amino-4-chlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32403G10 AS32403G5
ACTIAS32403G10EA 0 CHF
ACTIAS32403G10 ACTIAS32403G5
3-Amino-4-chlorbenzonitril
3-Amino-4-chlorbenzonitril
Formel: C₇H₅ClN₂
MDL Number: MFCD00017089
CAS-Nummer: 53312-79-1
EINECS: 258-471-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR