2,3-Dichlorpyrazin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS21431G100
AS21431G25
ACTIAS21431G100EA
0
CHF
ACTIAS21431G100
ACTIAS21431G25
2,3-Dichlorpyrazin
2,3-Dichloropyrazine
Formel:
C₄H₂Cl₂N₂ |
MDL Number:
MFCD00040964 CAS-Nummer: 4858-85-9 EINECS: 225-460-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...