3-Chlor-3-methyl-1-butin

Lieferant: ACTIVATE SCIENTIFIC

AS23502G25 AS23502G5
ACTIAS23502G25EA 291 CHF
ACTIAS23502G25 ACTIAS23502G5
3-Chlor-3-methyl-1-butin
3-Chlor-3-methyl-1-butin
Formel: C₅H₇Cl
MW: 102,56 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00190221
CAS-Nummer: 1111-97-3
EINECS: 214-188-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR