4-Nitrosalicylaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Hydroxy-4-nitrobenzaldehyde
AS36036G1
AS36036G5
ACTIAS36036G1EA
0
CHF
ACTIAS36036G1
ACTIAS36036G5
4-Nitrosalicylaldehyd
4-Nitrosalicylaldehyd
Formel:
C₇H₅NO₄ |
MDL Number:
MFCD00049373 CAS-Nummer: 2460-58-4 EINECS: 219-549-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...