3-Phenoxybenzylbromid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(Bromomethyl)-3-phenoxybenzene, 3-(Bromomethyl)diphenyl ether

AS36142G1 AS36142G5
ACTIAS36142G1EA 0 CHF
ACTIAS36142G1 ACTIAS36142G5
3-Phenoxybenzylbromid
3-Phenoxybenzylbromid
Formel: C₁₃H₁₁BrO
MW: 263,13 g/mol
MDL Number: MFCD01761658
CAS-Nummer: 51632-16-7
EINECS: 257-327-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR