2-Brombenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Bromophenyl)methanol

AS34467G10 AS34467G100
ACTIAS34467G10EA 0 CHF
ACTIAS34467G10 ACTIAS34467G100
2-Brombenzylalkohol
2-Brombenzylalkohol
Formel: C₇H₇BrO
MW: 187.04 g/mol
Siedepunkt: 255
Melting Pt: 80…82 °C
MDL Number: MFCD00004600
CAS-Nummer: 18982-54-2
EINECS: 242-719-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR