2-Amino-5-brom-3-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine
AS31741G1
AS31741G5
ACTIAS31741G1EA
0
CHF
ACTIAS31741G1
ACTIAS31741G5
2-Amino-5-brom-3-iodpyridin
2-Amino-5-brom-3-iodpyridin
Formel:
C₅H₄BrIN₂ MW: 298,91 g/mol |
MDL Number:
MFCD06659000 CAS-Nummer: 381233-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...