1-Brom-2-fluor-5-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS50346G1 AS50346G5
ACTIAS50346G1EA 0 CHF
ACTIAS50346G1 ACTIAS50346G5
1-Brom-2-fluor-5-nitrobenzol
1-Brom-2-fluor-5-nitrobenzol
Formel: C₆H₃BrFNO₂
MDL Number: MFCD00055433
CAS-Nummer: 701-45-1
EINECS: 211-855-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR