2-Amino-3-brom-5-fluorpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS30992G1
AS30992G5
ACTIAS30992G1EA
0
CHF
ACTIAS30992G1
ACTIAS30992G5
2-Amino-3-brom-5-fluorpyridin
2-Amino-3-bromo-5-fluoropyridine
Formel:
C₅H₄BrFN₂ |
MDL Number:
MFCD03092924 CAS-Nummer: 869557-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...