2-Brom-3-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS33849G1 AS33849G10 AS33849G5
ACTIAS33849G1EA 0 CHF
ACTIAS33849G1 ACTIAS33849G10 ACTIAS33849G5
2-Brom-3-(trifluormethyl)anilin
2-Brom-3-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
MW: 240.02 g/mol
MDL Number: MFCD01656033
CAS-Nummer: 58458-10-9
EINECS: 261-261-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR