2-Brom-3-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS31322G1 AS31322G5
ACTIAS31322G1EA 0 CHF
ACTIAS31322G1 ACTIAS31322G5
2-Brom-3-nitrobenzoesäure
2-Bromo-3-nitrobenzoic acid
Formel: C₇H₄BrNO₄
MW: 246,02 g/mol
MDL Number: MFCD00074899
CAS-Nummer: 573-54-6
EINECS: 209-356-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR