2-Brom-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS1501G100 AS1501G5
ACTIAS1501G100EA 0 CHF
ACTIAS1501G100 ACTIAS1501G5
2-Brom-m-toluylsäure
2-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
MDL Number: MFCD00079721
CAS-Nummer: 53663-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR