2,4-Dibrombenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS34250G1
AS34250G10
AS34250G5
ACTIAS34250G1EA
0
CHF
ACTIAS34250G1
ACTIAS34250G10
ACTIAS34250G5
2,4-Dibrombenzoesäure
2,4-Dibromobenzoic acid
Formel:
C₇H₄Br₂O₂ MW: 279,92 g/mol |
MDL Number:
MFCD00234253 CAS-Nummer: 611-00-7 EINECS: 210-245-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...