2-Amino-3-brom-5-chlorpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 3-Bromo-5-chloro-2-pyridinamine, 3-Bromo-5-chloro-2-pyridylamine, 3-Bromo-5-chloropyridin-2-amine

AS31900G1 AS31900G10 AS31900G5
ACTIAS31900G1EA 0 CHF
ACTIAS31900G1 ACTIAS31900G10 ACTIAS31900G5
2-Amino-3-brom-5-chlorpyridin
2-Amino-3-brom-5-chlorpyridin
Formel: C₅H₄BrClN₂
MDL Number: MFCD00955627
CAS-Nummer: 26163-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR