2-Brom-6-chlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS31119G1 AS31119G5
ACTIAS31119G1EA 0 CHF
ACTIAS31119G1 ACTIAS31119G5
2-Brom-6-chlorbenzonitril
2-Brom-6-chlorbenzonitril
Formel: C₇H₃BrClN
MW: 216,46 g/mol
MDL Number: MFCD06335184
CAS-Nummer: 6575-08-2
EINECS: 229-497-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR