2-Brombiphenyl (PBB Nr. 1)
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Bromobiphenyl, PBB No. 1, 2-Bromodiphenyl
AS24062G100
AS24062G25
ACTIAS24062G100EA
0
CHF
ACTIAS24062G100
ACTIAS24062G25
2-Brombiphenyl (PBB Nr. 1)
2-Brombiphenyl (PBB Nr. 1)
Formel:
C₁₂H₉Br MW: 233.11 g/mol Siedepunkt: 297 Melting Pt: 2 Dichte: 1.2766 g/cm³ (20 °C) |
MDL Number:
MFCD00000065 CAS-Nummer: 2052-07-5 EINECS: 218-141-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...