2,3-Dimethoxyphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2,3-Dimethoxybenzeneboronic acid, (2,3-Dimethoxyphenyl)metaboric acid, Dihydroxy(2,3-dimethoxyphenyl)borane
AS35026G1
AS35026G5
ACTIAS35026G1EA
0
CHF
ACTIAS35026G1
ACTIAS35026G5
2,3-Dimethoxyphenylboronsäure
2,3-Dimethoxyphenylboronsäure
Formel:
C₈H₁₁BO₄ MW: 181,98 g/mol |
MDL Number:
MFCD02683112 CAS-Nummer: 40972-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...