2-Cyanphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: (2-Cyanophenyl)metaboric acid, 2-Cyanobenzeneboronic acid, (2-Cyanophenyl)boronic acid, Dihydroxy(2-cyanophenyl)borane

AS51181G1 AS51181G5
ACTIAS51181G1EA 0 CHF
ACTIAS51181G1 ACTIAS51181G5
2-Cyanphenylboronsäure
2-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
MW: 146,94 g/mol
MDL Number: MFCD01632208
CAS-Nummer: 138642-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR