2,4-Dichlor-D-phenylalanin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: H-D-Phe(2,4-DiCl)-OH

AS31140G1 AS31140G10 AS31140G5
ACTIAS31140G1EA 0 CHF
ACTIAS31140G1 ACTIAS31140G10 ACTIAS31140G5
2,4-Dichlor-D-phenylalanin
2,4-Dichlor-D-phenylalanin
Formel: C₉H₉Cl₂NO₂
MW: 234.09 g/mol
MDL Number: MFCD01860873
CAS-Nummer: 114872-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR