2-Methyl-6-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS21408G1EA 85 CHF
ACTIAS21408G1 ACTIAS21408G5
2-Methyl-6-nitroanisol
2-Methyl-6-nitroanisol
Formel: C₈H₉NO₃
MDL Number: MFCD00661626
CAS-Nummer: 18102-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR