2-Chlor-5-fluorbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

AS35110G1 AS35110G10 AS35110G5
ACTIAS35110G1EA 0 CHF
ACTIAS35110G1 ACTIAS35110G10 ACTIAS35110G5
2-Chlor-5-fluorbenzotrifluorid
2-Chlor-5-fluorbenzotrifluorid
Formel: C₇H₃ClF₄
MW: 198,5500 g/mol
MDL Number: MFCD07782058
CAS-Nummer: 89634-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR