3-Brom-2-methylanisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Bromo-3-methoxy-2-methylbenzene

AS33772G1 AS33772G5
ACTIAS33772G1EA 0 CHF
ACTIAS33772G1 ACTIAS33772G5
3-Brom-2-methylanisol
3-Brom-2-methylanisol
Formel: C₈H₉BrO
MW: 201,0605 g/mol
MDL Number: MFCD11100991
CAS-Nummer: 31804-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR