3-Fluor-2-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS33845G1 AS33845G5
ACTIAS33845G1EA 0 CHF
ACTIAS33845G1 ACTIAS33845G5
3-Fluor-2-nitroanilin
3-Fluor-2-nitroanilin
Formel: C₆H₅FN₂O₂
MDL Number: MFCD07368750
CAS-Nummer: 567-63-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR