3-Fluor-N-methylbenzylamin

Lieferant: TCI EUROPE

TCIAF0805-1GEA 0 CHF
TCIAF0805-1G
3-Fluor-N-methylbenzylamin
3-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
MW: 139,17 g/mol
MDL Number: MFCD04623547
CAS-Nummer: 90389-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR