2-Amino-5-(trifluormethyl)benzonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Amino-3-cyanobenzotrifluoride, 2-Amino-5-trifluoromethylbenzonitrile
AS33669G1
AS33669G10
AS33669G5
ACTIAS33669G1EA
0
CHF
ACTIAS33669G1
ACTIAS33669G10
ACTIAS33669G5
2-Amino-5-(trifluormethyl)benzonitril
2-Amino-5-(trifluormethyl)benzonitril
Formel:
C₈H₅F₃N₂ MW: 186.1300 g/mol |
MDL Number:
MFCD03407465 CAS-Nummer: 6526-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...