1-(2-Bromethoxy)-2-ethoxybenzol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(2-Ethoxyphenoxy)ethyl bromide
AS33314G10
AS33314G5
ACTIAS33314G10EA
0
CHF
ACTIAS33314G10
ACTIAS33314G5
1-(2-Bromethoxy)-2-ethoxybenzol
1-(2-Bromoethoxy)-2-ethoxybenzene
Formel:
C₁₀H₁₃BrO₂ MW: 245,12 g/mol |
MDL Number:
MFCD02030483 CAS-Nummer: 3259-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...