4'-Brom-3'-fluoracetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-3-fluoro--acetophenone

AS8420G1 AS8420G5
ACTIAS8420G1EA 0 CHF
ACTIAS8420G1 ACTIAS8420G5
4'-Brom-3'-fluoracetophenon
4'-Brom-3'-fluoracetophenon
Formel: C₈H₆BrFO
MW: 217.04 g/mol
MDL Number: MFCD00466241
CAS-Nummer: 304445-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR