2-Fluor-6-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC

AS25056G1 AS25056G5
ACTIAS25056G1EA 46 CHF
ACTIAS25056G1 ACTIAS25056G5
2-Fluor-6-nitroanisol
2-Fluor-6-nitroanisol
Formel: C₇H₆FNO₃
MW: 171.1300 g/mol
MDL Number: MFCD06656241
CAS-Nummer: 484-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR