2-Fluor-6-methoxybenzylamin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(2-Fluoro-6-methoxyphenyl)methanamine
AS32265G1
AS32265G5
ACTIAS32265G1EA
0
CHF
ACTIAS32265G1
ACTIAS32265G5
2-Fluor-6-methoxybenzylamin
2-Fluor-6-methoxybenzylamin
Formel:
C₈H₁₀FNO MW: 155,1695 g/mol |
MDL Number:
MFCD00042292 CAS-Nummer: 150517-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...