5-Amino-2-fluor-4-nitrobenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoro-2-nitro-5-(trifluoromethyl)aniline

AS32293G1 AS32293G5
ACTIAS32293G1EA 0 CHF
ACTIAS32293G1 ACTIAS32293G5
5-Amino-2-fluor-4-nitrobenzotrifluorid
5-Amino-2-fluor-4-nitrobenzotrifluorid
Formel: C₇H₄F₄N₂O₂
MW: 224,11 g/mol
MDL Number: MFCD03094267
CAS-Nummer: 179062-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR