3-Amino-5-methylbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

AS32167G1 AS32167G5
ACTIAS32167G1EA 0 CHF
ACTIAS32167G1 ACTIAS32167G5
3-Amino-5-methylbenzotrifluorid
3-Amino-5-methylbenzotrifluorid
Formel: C₈H₈F₃N
MW: 175.1510 g/mol
MDL Number: MFCD01631580
CAS-Nummer: 96100-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR