5-Fluor-2-methylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid
AS50589G1
AS50589G5
ACTIAS50589G1EA
0
CHF
ACTIAS50589G1
ACTIAS50589G5
5-Fluor-2-methylphenylboronsäure
5-Fluor-2-methylphenylboronsäure
Formel:
C₇H₈BFO₂ MW: 153,95 g/mol |
MDL Number:
MFCD03095047 CAS-Nummer: 163517-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...