2-Fluor-5-methylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-5-methylbenzeneboronic acid, 2-Fluoro-5-methylboronic acid

AS34996G1 AS34996G10 AS34996G5
ACTIAS34996G1EA 0 CHF
ACTIAS34996G1 ACTIAS34996G10 ACTIAS34996G5
2-Fluor-5-methylphenylboronsäure
2-Fluor-5-methylphenylboronsäure
Formel: C₇H₈BFO₂
MDL Number: MFCD03428512
CAS-Nummer: 166328-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR