2,6-Dichlor-m-toluidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dichloro-3-methylaniline

AS22008G100 AS22008G25 AS22008G5
ACTIAS22008G100EA 0 CHF
ACTIAS22008G100 ACTIAS22008G25 ACTIAS22008G5
2,6-Dichlor-m-toluidin
2,6-Dichlor-m-toluidin
Formel: C₇H₇Cl₂N
MW: 176.0432 g/mol
MDL Number: MFCD00007676
CAS-Nummer: 64063-37-2
EINECS: 264-649-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR