5-Chlor-2-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-4-chlorobenzotrifluoride

AS32377G1 AS32377G10 AS32377G5
ACTIAS32377G1EA 0 CHF
ACTIAS32377G1 ACTIAS32377G10 ACTIAS32377G5
5-Chlor-2-(trifluormethyl)anilin
5-Chlor-2-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
MDL Number: MFCD08282771
CAS-Nummer: 445-14-7
EINECS: 207-153-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR