4-Brom-3-nitrophenol
Lieferant: ACTIVATE SCIENTIFIC
AS31005G1
AS31005G5
ACTIAS31005G1EA
0
CHF
ACTIAS31005G1
ACTIAS31005G5
4-Brom-3-nitrophenol
4-Brom-3-nitrophenol
Formel:
C₆H₄BrNO₃ MW: 218,0100 g/mol |
MDL Number:
MFCD11040194 CAS-Nummer: 78137-76-5 EINECS: 231-707-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...