2-Brom-5-nitrophenol
Lieferant: ACTIVATE SCIENTIFIC
AS51556G1
AS51556G5
ACTIAS51556G1EA
0
CHF
ACTIAS51556G1
ACTIAS51556G5
2-Brom-5-nitrophenol
2-Brom-5-nitrophenol
Formel:
C₆H₄BrNO₃ MW: 218,0100 g/mol |
MDL Number:
MFCD00466299 CAS-Nummer: 52427-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...