4-Isopropoxyphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Isopropoxybenzeneboronic acid
AS50846G100
AS50846G5
ACTIAS50846G100EA
0
CHF
ACTIAS50846G100
ACTIAS50846G5
4-Isopropoxyphenylboronsäure
4-Isopropoxyphenylboronsäure
Formel:
C₉H₁₃BO₃ MW: 180,01 g/mol |
MDL Number:
MFCD03427051 CAS-Nummer: 153624-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...