4-Bromzimtsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 3-(4-Bromophenyl)acrylic acid

AS23635G100 AS23635G25
ACTIAS23635G100EA 89 CHF
ACTIAS23635G100 ACTIAS23635G25
4-Bromzimtsäure
4-Bromzimtsäure
Formel: C₉H₇BrO₂
MW: 227,06 g/mol
MDL Number: MFCD00004394
CAS-Nummer: 1200-07-3
EINECS: 214-850-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR