2-Brom-3'-(trifluormethoxy)acetophenon
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Bromo-3'-trifluoromethoxyacetophenone, 3-(Trifluoromethoxy)phenacyl bromide
AS34436G1
AS34436G5
ACTIAS34436G1EA
0
CHF
ACTIAS34436G1
ACTIAS34436G5
2-Brom-3'-(trifluormethoxy)acetophenon
2-Brom-3'-(trifluormethoxy)acetophenon
Formel:
C₉H₆BrF₃O₂ MW: 283.0400 g/mol |
MDL Number:
MFCD03094425 CAS-Nummer: 237386-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...