4-(Aminomethyl)phenylboronsäure Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Aminomethylphenylboronic acid hydrochloride, 4-(Aminomethyl)benzeneboronic acid hydrochloride

AS50016G1 AS50016G5
ACTIAS50016G1EA 0 CHF
ACTIAS50016G1 ACTIAS50016G5
4-(Aminomethyl)phenylboronsäure Hydrochlorid
4-(Aminomethyl)phenylboronsäure Hydrochlorid
Formel: C₇H₁₁BClNO₂
MW: 187,43 g/mol
MDL Number: MFCD01632199
CAS-Nummer: 75705-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR