3-(Aminomethyl)phenylboronsäure Hydrochlorid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(3-(Aminomethyl)phenyl)boronic acid hydrochloride, 3-(Aminomethyl)benzeneboronic acid hydrochloride
AS50013G1
AS50013G5
ACTIAS50013G1EA
0
CHF
ACTIAS50013G1
ACTIAS50013G5
3-(Aminomethyl)phenylboronsäure Hydrochlorid
3-(Aminomethyl)phenylboronic acid hydrochloride
Formel:
C₇H₁₁BClNO₂ MW: 187,43 g/mol |
MDL Number:
MFCD02093050 CAS-Nummer: 352525-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...