2-Carboxy-4-fluorphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Borono-5-fluorobenzoic acid
AS33911G1
AS33911G5
ACTIAS33911G1EA
0
CHF
ACTIAS33911G1
ACTIAS33911G5
2-Carboxy-4-fluorphenylboronsäure
2-Carboxy-4-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ MW: 183.9296 g/mol |
MDL Number:
MFCD08235078 CAS-Nummer: 874290-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...