4-Carboxy-3-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Borono-2-fluorobenzoic acid, 4-Carboxy-3-fluorobenzeneboronic acid

AS50059G1 AS50059G5
ACTIAS50059G1EA 0 CHF
ACTIAS50059G1 ACTIAS50059G5
4-Carboxy-3-fluorphenylboronsäure
4-Carboxy-3-fluorphenylboronsäure
Formel: C₇H₆BFO₄
MW: 183,93 g/mol
MDL Number: MFCD01114671
CAS-Nummer: 120153-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR