2-Chlor-4-fluor-5-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene

AS22720G1 AS22720G5
ACTIAS22720G1EA 100 CHF
ACTIAS22720G1 ACTIAS22720G5
2-Chlor-4-fluor-5-nitroanisol
2-Chlor-4-fluor-5-nitroanisol
Formel: C₇H₅ClFNO₃
MW: 205,5700 g/mol
MDL Number: MFCD03425810
CAS-Nummer: 84478-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR