5-Carboxy-2-chlorphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Chloro-5-carboxyphenylboronic acid
AS34029G1
AS34029G10
AS34029G5
ACTIAS34029G1EA
0
CHF
ACTIAS34029G1
ACTIAS34029G10
ACTIAS34029G5
5-Carboxy-2-chlorphenylboronsäure
5-Carboxy-2-chlorphenylboronsäure
Formel:
C₇H₆BClO₄ MW: 200.3800 g/mol |
MDL Number:
MFCD08689491 CAS-Nummer: 913835-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...