3-Carboxy-4-chlorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Borono-2-chlorobenzoic acid

AS33921G1 AS33921G5
ACTIAS33921G1EA 0 CHF
ACTIAS33921G1 ACTIAS33921G5
3-Carboxy-4-chlorphenylboronsäure
3-Carboxy-4-chlorphenylboronsäure
Formel: C₇H₆BClO₄
MW: 200,3800 g/mol
MDL Number: MFCD06801689
CAS-Nummer: 913835-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR