4-Carboxy-3-chlorphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Carboxy-3-chlorobenzeneboronic acid
AS35118G1
AS35118G10
AS35118G5
ACTIAS35118G1EA
0
CHF
ACTIAS35118G1
ACTIAS35118G10
ACTIAS35118G5
4-Carboxy-3-chlorphenylboronsäure
4-Carboxy-3-chlorphenylboronsäure
Formel:
C₇H₆BClO₄ |
MDL Number:
MFCD06656268 CAS-Nummer: 136496-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...