2-Chlor-(4-methoxycarbonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-4-(methoxycarbonyl)phenylboronic acid, 2-Chloro-4-(methoxycarbonyl)benzeneboronic acid, (2-Chloro-4-(methoxycarbonyl)phenyl)boronic acid

AS34337G1 AS34337G10 AS34337G5
ACTIAS34337G1EA 0 CHF
ACTIAS34337G1 ACTIAS34337G10 ACTIAS34337G5
2-Chlor-(4-methoxycarbonyl)phenylboronsäure
2-Chlor-(4-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
MW: 214,4107 g/mol
MDL Number: MFCD08689530
CAS-Nummer: 603122-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR